missing translation for 'onlineSavingsMsg'
Learn More

5-Lipoxygenase-In-1, MedChemExpress

Product Code. 16462528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16462528 5 mg 5mg
16450329 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16462528 Supplier Medchem Express Supplier No. HYU00308
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 5-Lipoxygenase-In-1 is a 5-Lipoxygenase inhibitor extracted from patent EP 331232 A2, table 4, compound example 4.10.

5-Lipoxygenase-In-1 is a 5-Lipoxygenase inhibitor extracted from patent EP 331232 A2, table 4, compound example 4.10.

Specifications

Chemical Name or Material 5-Lipoxygenase-In-1
CAS 125235-15-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C23H28N4O2S
Quantity 5 mg
Solubility Information DMSO : 250 mg/mL (588.84 mM; Need ultrasonic)
SMILES O=C(C(C)(C)N1CC)N(C2=CC=C(N3CCN(C4=CC=C(O)C=C4)CC3)C=C2)C1=S
Molecular Weight (g/mol) 424.56
Formula Weight 424.56
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.