missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Lipoxygenase Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB110-58749AF647
This item is not returnable.
View return policy
Description
5-Lipoxygenase Polyclonal specifically detects 5-Lipoxygenase in Human, Mouse, Rat, Bovine, Rabbit samples. It is validated for Western Blot.
Specifications
| 5-Lipoxygenase | |
| Polyclonal | |
| Western Blot | |
| P09917 | |
| ALOX5 | |
| A synthetic peptide made to a C-terminal region within residues 600-674 of human 5-Lipoxygenase. [Swiss-Prot# P09917] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Bovine, Rabbit | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| 5-lipoxygenase, 5-LO, 5-LOX, 5LPG, arachidonate 5-lipoxygenase, arachidonic 5-lipoxygenase alpha-10 isoform, arachidonic 5-lipoxygenase delta-10-13 isoform, arachidonic 5-lipoxygenase delta-13 isoform, arachidonic 5-lipoxygenase delta-p10 isoform, arachidonic acid 5-lipoxygenase, EC 1.13.11, EC 1.13.11.34, leukotriene A4 synthase, LOG5, MGC163204 | |
| Rabbit | |
| 80 kDa | |
| 0.1 mL | |
| Cancer, Cholesterol Metabolism, Lipid and Metabolism, Neuroscience, Phospho Specific, Signal Transduction | |
| 240 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction