missing translation for 'onlineSavingsMsg'
Learn More

5-Keto-D-gluconic acid potassium salt, 98%

Product Code. 15428479
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15428479 1 g 1g
15438479 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15428479 Supplier Thermo Scientific Alfa Aesar Supplier No. J66916.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5447-60-9 | C6H10KO7 | 233.237 g/mol

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in water 50mg/ml

Notes
Store at -20°C. Incompatible with heat, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C6H10KO7
Molecular Weight (g/mol) 233.237
MDL Number MFCD00069562
InChI Key BJBNFUCYWPVFKM-YMDUGQBDSA-N
Synonym potassium 5-ketogluconate
PubChem CID 131855000
IUPAC Name potassium;(2R,3S,4S)-2,3,4,6-tetrahydroxy-5-oxohexanoic acid
SMILES C(C(=O)C(C(C(C(=O)O)O)O)O)O.[K]

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water 50mg/ml
Formula Weight 232.23
Percent Purity 0.98
Physical Form Powder
Chemical Name or Material 5-Keto-D-gluconic acid potassium salt
compliance-icons
Product Identifier
  • 5-Keto-D-gluconic acid potassium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-Keto-D-gluconic acid potassium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.