missing translation for 'onlineSavingsMsg'
Learn More

5-Isoxazol-5-ylthiophene-2-sulfonyl chloride, Tech., Thermo Scientific™

Product Code. 10167712
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10167712 1 g 1g
10157712 250 mg 250mg
10664575 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10167712 Supplier Thermo Scientific Maybridge Supplier No. CC16003DA
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 551930-53-1
Molecular Formula C7H4ClNO3S2
Molecular Weight (g/mol) 249.68
InChI Key SSGKBJJYLTYNQD-UHFFFAOYSA-N
Synonym 5-5-isoxazyl thiophene-2-sulfonyl chloride, 5-isoxazol-5-ylthiophene-2-sulfonyl chloride, 5-1,2-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonyl chloride, 5-isoxazol-5-yl thiophene-2-sulfonyl chloride, chloro 5-isoxazol-5-yl 2-thienyl sulfone, ssgkbjjyltynqd-uhfffaoysa-n, 5-5-isoxazyl thiophene-2-sulphonyl chloride, 5-5-isoxazolyl-2-thiophenesulfonoyl chloride, 5-5-isoxazolyl thiophene-2-sulfonyl chloride
PubChem CID 2795330

Specifications

Quantity 1 g
Formula Weight 249.7
Percent Purity Tech.
Grade Technical
Chemical Name or Material 5-Isoxazol-5-ylthiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.