missing translation for 'onlineSavingsMsg'
Learn More

5-Isopropylbarbituric acid, 98%, Thermo Scientific™

Product Code. 11337096
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7391-69-7
Molecular Formula C7H10N2O3
Molecular Weight (g/mol) 170.168
MDL Number MFCD00051832
InChI Key GFPIGNBQTXNNAG-UHFFFAOYSA-N
Synonym 5-isopropylbarbituric acid, 2,4,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-isopropylpyrimidine-2,4,6 1h,3h,5h-trione, 5-isopropyl barbituric acid, barbituric acid, 5-isopropyl, barbituric acid, 5-ipr, 2,6 1h,3h,5h-pyrimidinetrione, 5-1-methylethyl, 5-1-methylethyl pyrimidine-2,4,6 1h,3h,5h-trione
PubChem CID 81874
IUPAC Name 5-propan-2-yl-1,3-diazinane-2,4,6-trione
SMILES CC(C)C1C(=O)NC(=O)NC1=O

Specifications

Melting Point 213°C to 216°C
Beilstein 149805
Formula Weight 170.17
Percent Purity 98%
Quantity 5 g
Chemical Name or Material 5-Isopropylbarbituric acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'