missing translation for 'onlineSavingsMsg'
Learn More

5-Iodovanillic Acid, TRC

Product Code. 30392535
Change view
Click to view available options
Quantity:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392535 10 g
30395930 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30392535 Supplier Toronto Research Chemicals Supplier No. TRCI68551310G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 98554-57-5
Molecular Formula C8H7IO4
Molecular Weight (g/mol) 294.04
IUPAC Name 4-hydroxy-3-iodo-5-methoxybenzoic acid
SMILES COc1cc(cc(I)c1O)C(=O)O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C8H7IO4/c1-13-6-3-4(8(11)12)2-5(9)7(6)10/h2-3,10H,1H3,(H,11,12)
Formula Weight 293.9389
Chemical Name or Material 5-Iodovanillic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.