missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Iodouracil, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 5-Iodouracil |
| CAS | 696-07-1 |
| Molecular Formula | C4H3IN2O2 |
| Quantity | 10 g |
| Synonym | 5-Iodo-2,4(1H,3H)-pyrimidinedione, 5-Iodo-2,4-dihydroxypyrimidine, 5-Iodo-2,4-pyrimidinedione, 5-Iodopyrimidine-2,4-diol, 5-Iodouracil, NSC 57848 |
| InChI Formula | InChI=1S/C4H3IN2O2/c5-2-1-6-4(9)7-3(2)8/h1H,(H2,6,7,8,9) |
| SMILES | IC1=CNC(=O)NC1=O |
| Molecular Weight (g/mol) | 237.98 |
| Formula Weight | 237.9239 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction