missing translation for 'onlineSavingsMsg'
Learn More

5-Iodotubercidin, MedChemExpress

Product Code. 16479087
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
25 mg
50 mg
Unit Size:
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16479087

Brand: Medchem Express HY15424

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Iodotubercidin (NSC 113939), an ATP mimetic, is a potent adenosine kinase inhibitor with an IC50 of 26 nM. 5-Iodotubercidin (NSC 113939) initiates glycogen synthesis in isolated hepatocytes by causing inactivation of phosphorylase and activation of glycogen synthase. 5-Iodotubercidin (NSC 113939) also inhibits CK1, insulin receptor tyrosine kinase, phosphorylase kinase, PKA, CK2, PKC and Haspin.

5-Iodotubercidin (NSC 113939), an ATP mimetic, is a potent adenosine kinase inhibitor with an IC50 of 26 nM. 5-Iodotubercidin (NSC 113939) initiates glycogen synthesis in isolated hepatocytes by causing inactivation of phosphorylase and activation of glycogen synthase. 5-Iodotubercidin (NSC 113939) also inhibits CK1, insulin receptor tyrosine kinase, phosphorylase kinase, PKA, CK2, PKC and Haspin.

Specifications

Chemical Name or Material 5-Iodotubercidin
CAS 24386-93-4
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C11H13IN4O4
Quantity 5 mg
Synonym NSC 113939 5-ITu
Solubility Information DMSO : ≥ 49 mg/mL (124.95 mM)
SMILES NC1=C2C(N([C@H]3[C@H](O)[C@H](O)[C@@H](CO)O3)C=C2I)=NC=N1
Molecular Weight (g/mol) 392.15
Formula Weight 392.15
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.