missing translation for 'onlineSavingsMsg'
Learn More

5-Iodotoluic Acid, TRC

Product Code. 30397107
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397107 100 mg 100mg
30400153 250 mg -
30403100 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30397107 Supplier Toronto Research Chemicals Supplier No. TRCI708645100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133232-58-3
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.04
Synonym 4-Iodo-2-methylbenzoic Acid
IUPAC Name 4-iodo-2-methylbenzoic acid
SMILES Cc1cc(I)ccc1C(=O)O

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H7IO2/c1-5-4-6(9)2-3-7(5)8(10)11/h2-4H,1H3,(H,10,11)
Formula Weight 261.9491
Chemical Name or Material 5-Iodotoluic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.