missing translation for 'onlineSavingsMsg'
Learn More

5-Iodosalicylic Acid 97%, ACROS Organics™

Product Code. 10272652 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10272652 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10272652 Supplier Thermo Scientific Acros Supplier No. 122420050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 119-30-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 196°C to 201°C
Color Beige to White
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C7H5IO3
MDL Number MFCD00002458
Quantity 5 g
Beilstein 10, 112
Synonym 5-iodosalicylic acid, benzoic acid, 2-hydroxy-5-iodo, salicylic acid, 5-iodo, 5-iodo-2-hydroxybenzoic acid, 2-hydroxy-5-iodo-benzoic acid, rarechem al be 0824, labotest-bb lt00454718, 5-jod-salicylsaure, 5-iodosalicylicacid, 5-iodo-salicylicaci
InChI Key SWDNKOFGNPGRPI-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1I)C(=O)O)O
IUPAC Name 2-hydroxy-5-iodobenzoic acid
Molecular Weight (g/mol) 264.018
PubChem CID 8388
Formula Weight 264.01
Percent Purity ≥96.0%
Physical Form Powder
Chemical Name or Material 5-Iodosalicylic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • 5-Iodosalicylic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.