missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-m-xylene, 99%, Thermo Scientific Chemicals

Product Code. 10511082
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22445-41-6
Molecular Formula C8H9I
Molecular Weight (g/mol) 232.06
MDL Number MFCD00060659
InChI Key ZLMKEENUYIUKKC-UHFFFAOYSA-N
Synonym 5-iodo-m-xylene, 1,3-dimethyl-5-iodobenzene, benzene, 1-iodo-3,5-dimethyl, 3,5-dimethyliodobenzene, pubchem3872, acmc-1ccdf, 3,5-dimethyl-1-iodobenzene, ksc422q9l, 1-iodo-3,5-dimethylbenzene, #
PubChem CID 140924
IUPAC Name 1-iodo-3,5-dimethylbenzene
SMILES CC1=CC(I)=CC(C)=C1

Specifications

CAS Min % 98.5
Color Yellow
CAS Max % 100.0
Density 1.6000g/mL
Boiling Point 92°C to 94°C (3.0 mmHg)
Quantity 5 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5930 to 1.595
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.6
Formula Weight 232.07
Percent Purity 99%
Chemical Name or Material 5-Iodo-m-xylene, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'