missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-pyrrolidin-1ylpyridine, Thermo Scientific™

Product Code. 10361130
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10361130 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10361130 Supplier Thermo Scientific Maybridge Supplier No. MAY00164DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 494771-62-9
Packaging Amber Glass Bottle
Molecular Formula C9H11IN2
Quantity 1 g
Synonym 5-iodo-2-pyrrolidin-1-yl pyridine, 5-iodo-2-pyrrolidin-1ylpyridine, 5-iodo-2-1-pyrrolidinyl pyridine, acmc-1aqgd, 5-iodo-2-1-pyrrolidino pyridine
InChI Key PAGIQNSDEQQUIO-UHFFFAOYSA-N
SMILES C1CCN(C1)C2=NC=C(C=C2)I
IUPAC Name 5-iodo-2-pyrrolidin-1-ylpyridine
Molecular Weight (g/mol) 274.105
PubChem CID 21914402
Formula Weight 274.1
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-pyrrolidin-1ylpyridine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.