missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-methylaniline, 98%, Thermo Scientific™

Product Code. 11361897
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11361897 1 g 1g
11371897 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11361897 Supplier Thermo Scientific Alfa Aesar Supplier No. L07404.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 83863-33-6
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.05
MDL Number MFCD00051531
InChI Key IOEHXNCBPIBDBZ-UHFFFAOYSA-N
Synonym 2-amino-4-iodotoluene, 5-iodo-o-toluidine, 5-iodo-2-methyl-phenylamine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl, 5-iodo-2-methyl-aniline, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 2-methyl-5-iodoaniline, #
PubChem CID 522802
IUPAC Name 5-iodo-2-methylaniline
SMILES CC1=CC=C(I)C=C1N

Specifications

Melting Point 46°C to 49°C
Quantity 1 g
UN Number UN2811
Beilstein 2078769
Sensitivity Light sensitive
Formula Weight 233.05
Percent Purity 98%
Chemical Name or Material 5-Iodo-2-methylaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.