missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-methylaniline, 97%, Thermo Scientific™

Product Code. 11935231 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11935231 5 g 5g
11925231 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11935231 Supplier Thermo Scientific Acros Supplier No. 442430050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 83863-33-6
Molecular Formula C7H8IN
Molecular Weight (g/mol) 233.05
MDL Number MFCD00051531
InChI Key IOEHXNCBPIBDBZ-UHFFFAOYSA-N
Synonym 2-amino-4-iodotoluene, 5-iodo-o-toluidine, 5-iodo-2-methyl-phenylamine, benzenamine, 5-iodo-2-methyl, benzenamine,5-iodo-2-methyl, 5-iodo-2-methyl-aniline, 5-iodo-2-methylbenzenamine, 5-iodo-2-methylphenylamine, 2-methyl-5-iodoaniline, #
PubChem CID 522802
IUPAC Name 5-iodo-2-methylaniline
SMILES CC1=CC=C(I)C=C1N

Specifications

Melting Point 48°C to 50°C
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 233.05
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-methylaniline
compliance-icons compliance-icons
Product Identifier
  • 5-Iodo-2-methylaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.