missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-methoxybenzaldehyde, 97%

Product Code. 15462729
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15462729 5 g 5g
15452729 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15462729 Supplier Thermo Scientific Alfa Aesar Supplier No. H66320.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 42298-41-9 | C8H7IO2 | 262.05 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42298-41-9
Molecular Formula C8H7IO2
Molecular Weight (g/mol) 262.05
MDL Number MFCD04128999
InChI Key PIRKMHAREMCDPZ-UHFFFAOYSA-N
Synonym 2-methoxy-5-iodobenzaldehyde, 5-iodo-2-methoxy-benzaldehyde, pubchem1445, acmc-20am2z, benzaldehyde,5-iodo-2-methoxy, 5-iodo-2-methoxybenzaldehyde
PubChem CID 262240
IUPAC Name 5-iodo-2-methoxybenzaldehyde
SMILES COC1=CC=C(I)C=C1C=O

Specifications

Melting Point 140°C to 146°C
Color White to Yellow
Quantity 5 g
Sensitivity Air and light sensitive
Formula Weight 262.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Iodo-2-methoxybenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.