missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-methoxyaniline, 97%

Product Code. 11487236
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11487236

Brand: Thermo Scientific Alfa Aesar B25557.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 77770-09-3 | C7H8INO | 249.051 g/mol

5-Iodo-2-methoxyaniline is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-2-methoxyaniline is used as a pharmaceutical intermediate.

Solubility
Sparingly soluble in water 0.87 g/L @ 25°C.

Notes
Light sensitive. Store in dark. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77770-09-3
Molecular Formula C7H8INO
Molecular Weight (g/mol) 249.051
MDL Number MFCD03095407
InChI Key KCAGMESWEMVFON-UHFFFAOYSA-N
Synonym 2-amino-4-iodoanisole, 5-iodo-o-anisidine, 2-methoxy-5-iodoaniline, benzenamine, 5-iodo-2-methoxy, 4-methoxy-3-amino-1-iodobenzene, 5-iodo-2-methoxybenzenamine, 5-iodo-2-methoxy-phenylamine, 2-amino-4-iodoanisole 5-iodo-2-methoxyaniline
PubChem CID 12905382
IUPAC Name 5-iodo-2-methoxyaniline
SMILES COC1=C(C=C(C=C1)I)N

Specifications

Melting Point 86°C to 87°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Sparingly soluble in water 0.87g/L @ 25°C.
Formula Weight 249.05
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-methoxyaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.