missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-2-furaldehyde, 97%

Product Code. 11355485
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11355485 1 g 1g
11365485 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11355485 Supplier Thermo Scientific Alfa Aesar Supplier No. L19670.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2689-65-8 | C5H3IO2 | 221.981 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2689-65-8
Molecular Formula C5H3IO2
Molecular Weight (g/mol) 221.981
MDL Number MFCD00159503
InChI Key QPGPCPKDVSPJAY-UHFFFAOYSA-N
Synonym 5-iodo-2-furaldehyde, 5-iodofurfural, 5-iodo-2-furancarboxaldehyde, 5-iodo-2-furaldehdye, 2-furancarboxaldehyde, 5-iodo, 5-iodofuraldehyde, acmc-209gtv, 5-iodofurancarboxaldehyde, 5-iodanylfuran-2-carbaldehyde
PubChem CID 693264
IUPAC Name 5-iodofuran-2-carbaldehyde
SMILES C1=C(OC(=C1)I)C=O

Specifications

Melting Point 126°C to 127°C
Quantity 1 g
Beilstein 108406
Sensitivity Air and light sensitive
Formula Weight 221.98
Percent Purity 97%
Chemical Name or Material 5-Iodo-2-furaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.