missing translation for 'onlineSavingsMsg'
Learn More

(+)-5-Iodo-2'-deoxyuridine, 99%

Product Code. 10054043 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10054043 25 g Glass bottle 25g
10225070 1 g Glass bottle 1g
10649843 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10054043 Supplier Thermo Scientific Acros Supplier No. 122350250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 54-42-2 | C9H11IN2O5 | 354.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-42-2
Molecular Formula C9H11IN2O5
Molecular Weight (g/mol) 354.10
MDL Number MFCD00134656
InChI Key XQFRJNBWHJMXHO-RRKCRQDMSA-N
Synonym idoxuridine, 5-iodo-2'-deoxyuridine, idoxuridin, 5-iododeoxyuridine, iododeoxyridine, iodoxuridine, iudr, joddeoxiuridin, idoxene, allergan 211
PubChem CID 5905
ChEBI CHEBI:147675
SMILES OC[C@H]1O[C@H](C[C@@H]1O)N1C=C(I)C(=O)NC1=O

Specifications

CAS Min % 98.5
CAS Max % 100
Melting Point 155.0°C to 180.0°C
Color White to Beige
Loss on Drying 1% max. (1 g, 60°C) (vacuum)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 4931
Specific Rotation Condition + 280.00 (20.00°C c=1,1M NaOH)
Specific Rotation + 280.00
Solubility Information Solubility in water: 1.6g/L (20°C). Other solubilities: 2.0g/L hcl 0.2n- 74.0g/L naoh 0.2n (25°C),4.4g/L methanol- 2.6g/L alcohol (25°C),0.014g/L ether- 0.003g/L chloroform (25°C),1.6g/L acetone- 1.8g/L ethyl acetate (25°C),5.7
IUPAC Name 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-5-iodopyrimidine-2,4-dione
Formula Weight 354.09
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material (+)-5-Iodo-2′-deoxyuridine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • (+)-5-Iodo-2'-deoxyuridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H361d-Suspected of damaging the unborn child.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.