missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-1,2,3-trimethoxybenzene, 97%

Product Code. p-7047035
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11358396

Brand: Thermo Scientific Alfa Aesar L17414.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25245-29-8 | C9H11IO3 | 294.09 g/mol

5-Iodo-1,2,3-trimethoxybenzene is used as a primary and secondary intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-1,2,3-trimethoxybenzene is used as a primary and secondary intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25245-29-8
Molecular Formula C9H11IO3
Molecular Weight (g/mol) 294.09
MDL Number MFCD01318153
InChI Key IWPMQXOVTMABLF-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyiodobenzene, benzene, 5-iodo-1,2,3-trimethoxy, acmc-1cfcc, 3,4,5-trimethoxy-iodobenzene, 3,4,5-trimethoxyphenyl iodide, 3,4,5-trimethoxy-iodo-benzene, 1-iodo-3,4,5-trimethoxybenzene, 1,2,3-trimethoxy-5-iodobenzene, 5-iodo-1,2,3-trimethoxy-benzene, 5-iodanyl-1,2,3-trimethoxy-benzene
PubChem CID 298133
IUPAC Name 5-iodo-1,2,3-trimethoxybenzene
SMILES COC1=CC(I)=CC(OC)=C1OC

Specifications

Melting Point 85°C to 87°C
Quantity 1 g
Beilstein 1529690
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 294.09
Percent Purity ≥98%
Chemical Name or Material 5-Iodo-1,2,3-trimethoxybenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.