missing translation for 'onlineSavingsMsg'
Learn More

5-Iodo-1,2,3-trimethoxybenzene, 97%

Product Code. 11358396
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11358396 1 g 1g
11368396 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358396 Supplier Thermo Scientific Alfa Aesar Supplier No. L17414.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25245-29-8 | C9H11IO3 | 294.09 g/mol

5-Iodo-1,2,3-trimethoxybenzene is used as a primary and secondary intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Iodo-1,2,3-trimethoxybenzene is used as a primary and secondary intermediate in organic synthesis.

Solubility
Insoluble in water.

Notes
It is sensitive to light. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25245-29-8
Molecular Formula C9H11IO3
Molecular Weight (g/mol) 294.09
MDL Number MFCD01318153
InChI Key IWPMQXOVTMABLF-UHFFFAOYSA-N
Synonym 3,4,5-trimethoxyiodobenzene, benzene, 5-iodo-1,2,3-trimethoxy, acmc-1cfcc, 3,4,5-trimethoxy-iodobenzene, 3,4,5-trimethoxyphenyl iodide, 3,4,5-trimethoxy-iodo-benzene, 1-iodo-3,4,5-trimethoxybenzene, 1,2,3-trimethoxy-5-iodobenzene, 5-iodo-1,2,3-trimethoxy-benzene, 5-iodanyl-1,2,3-trimethoxy-benzene
PubChem CID 298133
IUPAC Name 5-iodo-1,2,3-trimethoxybenzene
SMILES COC1=CC(I)=CC(OC)=C1OC

Specifications

Melting Point 85°C to 87°C
Quantity 1 g
Beilstein 1529690
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 294.09
Percent Purity ≥98%
Chemical Name or Material 5-Iodo-1,2,3-trimethoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.