missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 5-InoAz
Description
5-InoAz is a metabolic probe for labeling the mycobacterium cell envelope. Specifically designed as an inositol analog to incorportate into cell envelope glycolipids (phosphatidylinositol mannoside (PIM), PIM-anchored lipomannan (LM) and lipoarabinomannan (LAM)). In combination with bioorthogonal chemistry, it provides a valuable tool for investigating PIM, LM, and LAM biosynthesis, transport, and dynamics in diverse mycobacterial organisms.
Specifications
Specifications
| Chemical Name or Material | 5-Azido-5-deoxy-myo-inositol |
| CAS | 1218932-97-8 |
| Quantity | 50 mg |
| Target | Glycobiology Probes |
| Molecular Formula | C6H11N3O5 |
| Purity | 0.9 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?