missing translation for 'onlineSavingsMsg'
Learn More

5-Imidazolecarboxaldehyde, TRC

Product Code. 30392484
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392484 10 g
30402184 1 g
30394537 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392484 Supplier Toronto Research Chemicals Supplier No. TRCI35022010G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3034-50-2
Molecular Formula C4H4N2O
Molecular Weight (g/mol) 96.09
Synonym 1H-Imidazole-5-carboxaldehyde, 1H-Imidazole-4-carboxaldehyde, Imidazole-4(or 5)-carboxaldehyde, Imidazole-4-carboxaldehyde (8CI), 1H-Imidazol-4-carboxaldehyde, 1H-Imidazol-4-ylcarboxaldehyde, 1H-Imidazol-5-carboxaldehyde, 1H-Imidazole-4-carboxaldehyde, 3H-Imidazole-4-carboxaldehyde, 4(5)-Imidazolecarboxaldehyde, 4-Formylimidazole, 5-Imidazolecarboxaldehyde, NSC 400521
SMILES O=Cc1c[nH]cn1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C4H4N2O/c7-2-4-1-5-3-6-4/h1-3H,(H,5,6)
Formula Weight 96.0324
Chemical Name or Material 5-Imidazolecarboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.