missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxytryptophol, MedChemExpress

Product Code. 16475457
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16475457 50 mg 50mg
16420489 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16475457 Supplier Medchem Express Supplier No. HYW041019

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Hydroxytryptophol is a mammalian serotonin metabolite, acting as a marker of acute alcohol consumption.

5-Hydroxytryptophol is a mammalian serotonin metabolite, acting as a marker of acute alcohol consumption.

Specifications

Chemical Name or Material 5-Hydroxytryptophol
CAS 154-02-9
Color Yellow
Purity Grade Notes Research
Molecular Formula C10H11NO2
Quantity 50 mg
Solubility Information DMSO : 125 mg/mL (705.42 mM; Need ultrasonic)
SMILES OC1=CC2=C(NC=C2CCO)C=C1
Molecular Weight (g/mol) 177.2
Formula Weight 177.2
Percent Purity 95.8%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.