missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxytryptophan, MedChemExpress

Product Code. 16488868
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16488868 10 mg 10mg
16484757 50 mg 50mg
16408996 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488868 Supplier Medchem Express Supplier No. HYN0122

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 5-Hydroxytryptophan, a tryptophan metabolite, is a direct 5-hydroxytryptamine (5-HT) precursor and an L-aromatic amino acid decarboxylase substrate. .

5-Hydroxytryptophan, a tryptophan metabolite, is a direct 5-hydroxytryptamine (5-HT) precursor and an L-aromatic amino acid decarboxylase substrate. .

Specifications

Chemical Name or Material 5-Hydroxytryptophan
CAS 56-69-9
Color Off-White
Purity Grade Notes Research
Molecular Formula C11H12N2O3
Synonym 5-HTP DL-5-Hydroxytryptophan
Solubility Information H2O : 12.5 mg/mL (56.76 mM; Need ultrasonic)
SMILES NC(CC1=CNC2=CC=C(O)C=C12)C(O)=O
Molecular Weight (g/mol) 220.22
Quantity 10 mg
Formula Weight 220.22
Percent Purity 98.99%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.