missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxytetrahydro-2H-pyran-2-one, TRC

Product Code. 30406763
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30406763 50 mg
30391838 10 mg
30389858 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30406763 Supplier Toronto Research Chemicals Supplier No. TRCH36917550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 33691-73-5
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.12
IUPAC Name 5-hydroxyoxan-2-one
SMILES OC1CCC(=O)OC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C5H8O3/c6-4-1-2-5(7)8-3-4/h4,6H,1-3H2
Formula Weight 116.0473
Chemical Name or Material 5-Hydroxytetrahydro-2H-pyran-2-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.