missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxypyridine-2-carboxylic acid, 98+%

Product Code. 15431379
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431379 1 g 1g
15441379 5 g 5g
15451379 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431379 Supplier Thermo Scientific Alfa Aesar Supplier No. H64265.03
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 15069-92-8 | C6H5NO3 | 139.11 g/mol

Useful synthetic intermediate. Also intermediate for pharmaceutical application.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful synthetic intermediate. Also intermediate for pharmaceutical application.

Solubility
Soluble in acetone, dimethyl sulfoxide , ethanol and methanol.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15069-92-8
Molecular Formula C6H5NO3
Molecular Weight (g/mol) 139.11
MDL Number MFCD00661309
InChI Key CGRRUFNHHQCLDZ-UHFFFAOYSA-N
Synonym 5-hydroxypicolinic acid, 5-hydroxy-2-pyridinecarboxylic acid, 5-hydroxy-pyridine-2-carboxylic acid, 6-carboxy-3-pyridinol, 2-pyridinecarboxylic acid, 5-hydroxy, 5-hydroxy-2-pyridine carboxylic acid, pubchem10094, 5-hydroxypicolinic acidn, acmc-209d4x, ksc171o9d
PubChem CID 268740
IUPAC Name 5-hydroxypyridine-2-carboxylic acid
SMILES C1=CC(=NC=C1O)C(=O)O

Specifications

Melting Point 260°C to 264°C
Quantity 1 g
Beilstein 116009
Solubility Information Soluble in acetone,dimethyl sulfoxide ,ethanol and methanol.
Formula Weight 139.11
Percent Purity ≥98%
Physical Form Solid
Chemical Name or Material 5-Hydroxypyridine-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.