missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxypyrazine-2-carboxylic Acid, TRC

Product Code. 17993305
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17993305 1 g 1g
17935005 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17993305 Supplier Toronto Research Chemicals Supplier No. TRCH9526751G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34604-60-9
Molecular Formula C5 H4 N2 O3
Molecular Weight (g/mol) 140.1
Synonym 4,5-Dihydro-5-oxopyrazinecarboxylic Acid, 5-Hydroxypyrazinecarboxylic Acid, 5-Hydroxypyrazinoic acid, 5-Oxo-4,5-dihydropyrazine-2-carboxylic Acid
IUPAC Name 5-hydroxypyrazine-2-carboxylic acid
SMILES OC(=O)c1cnc(O)cn1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C5H4N2O3/c8-4-2-6-3(1-7-4)5(9)10/h1-2H,(H,7,8)(H,9,10)
Formula Weight 140.0222
Chemical Name or Material 5-Hydroxypyrazine-2-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.