missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxynitrofen, TRC

Product Code. 30400939
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400939 5 g 5g
30402590 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30400939 Supplier Toronto Research Chemicals Supplier No. TRCH9493055G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63987-04-2
Molecular Formula C12H7Cl2NO4
Molecular Weight (g/mol) 300.09
Synonym 2,4-Dichloro-5-hydroxyphenyl 4-nitrophenyl Ether, 2,4-Dichloro-5-(4-nitrophenoxy)phenol
IUPAC Name 2,4-dichloro-5-(4-nitrophenoxy)phenol
SMILES Oc1cc(Oc2ccc(cc2)[N+](=O)[O-])c(Cl)cc1Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C12H7Cl2NO4/c13-9-5-10(14)12(6-11(9)16)19-8-3-1-7(2-4-8)15(17)18/h1-6,16H
Formula Weight 298.9752
Chemical Name or Material 5-Hydroxynitrofen
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.