missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxymethylfurfural, MedChemExpress

Product Code. 16425467
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16425467 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16425467 Supplier Medchem Express Supplier No. HYY0051
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5-Hydroxymethylfurfural (2-Hydroxymethyl-5-furfural), derived from lignocellulosic biomass, inhibits yeast growth and fermentation as stressors.

5-Hydroxymethylfurfural (2-Hydroxymethyl-5-furfural), derived from lignocellulosic biomass, inhibits yeast growth and fermentation as stressors.

Specifications

Chemical Name or Material 5-Hydroxymethylfurfural
CAS 67-47-0
Color Brown
Purity Grade Notes Research
Molecular Formula C6H6O3
Quantity 50 mg
Synonym 2-Hydroxymethyl-5-furfural 2-Formyl-5-hydroxymethylfuran
Solubility Information DMSO : ≥ 50 mg/mL (396.48 mM)
SMILES O=CC1=CC=C(CO)O1
Molecular Weight (g/mol) 126.11
Formula Weight 126.11
Percent Purity 97.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.