missing translation for 'onlineSavingsMsg'
Learn More

5-(Hydroxymethyl)uracil, 98%

Product Code. 11331377
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11331377 1 g 1g
11341377 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11331377 Supplier Thermo Scientific Alfa Aesar Supplier No. L01682.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4433-40-3 | C5H6N2O3 | 142.11 g/mol

5-(Hydroxymethyl)uracil serves as a stable major oxidative modification of thymine formed through the action of ionizing radiation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-(Hydroxymethyl)uracil serves as a stable major oxidative modification of thymine formed through the action of ionizing radiation.

Solubility
Soluble in water. (50 g/L ) at 20°C, DMSO, dimethyl formamide or 100% ethanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4433-40-3
Molecular Formula C5H6N2O3
Molecular Weight (g/mol) 142.11
MDL Number MFCD00056024
InChI Key JDBGXEHEIRGOBU-UHFFFAOYSA-N
Synonym 5-hydroxymethyluracil, 5-hydroxymethyl uracil, 5-hydroxymethyl pyrimidine-2,4 1h,3h-dione, 5-oxymethyluracil, 4-methyl-5-oxyuracil, uracil, 5-hydroxymethyl, 2,4 1h,3h-pyrimidinedione, 5-hydroxymethyl, thymine, alpha-hydroxy, 5-hydroxymethyl-2,4 1h,3h-pyrimidinedione
PubChem CID 78168
ChEBI CHEBI:16964
IUPAC Name 5-(hydroxymethyl)-1H-pyrimidine-2,4-dione
SMILES OCC1=CNC(=O)NC1=O

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 125482
Solubility Information Soluble in water. (50g/L ) at 20°C,DMSO,dimethyl formamide or 100% ethanol.
Formula Weight 142.12
Percent Purity 98%
Chemical Name or Material 5-(Hydroxymethyl)uracil

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.