missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxymethyl-2-furancarboxylic Acid-13C6, TRC

Product Code. 30396953
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30396953 10 mg
30406362 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30396953 Supplier Toronto Research Chemicals Supplier No. TRCH94702210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula 13C6H6O4
Molecular Weight (g/mol) 148.07
Synonym Sumikis' Acid-13C6, 5-(Hydroxymethyl)-2-furoic Acid13C6, NSC 40739-13C6
SMILES O[13C]([13C]1=[13C]([H])[13C]([H])=[13C]([13C]([H])([H])O)O1)=O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C6H6O4/c7-3-4-1-2-5(10-4)6(8)9/h1-2,7H,3H2,(H,8,9)/i1+1,2+1,3+1,4+1,5+1,6+1
Chemical Name or Material 5-Hydroxymethyl-2-furancarboxylic Acid-13C6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.