missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole, 98+%

Product Code. 11394324
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394324 1 g 1g
11304334 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394324 Supplier Thermo Scientific Alfa Aesar Supplier No. L00858.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1953-54-4 | C8H7NO | 133.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1953-54-4
Molecular Formula C8H7NO
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005677
InChI Key LMIQERWZRIFWNZ-UHFFFAOYSA-N
Synonym 5-hydroxyindole, indol-5-ol, 5-hydroxy-1h-indole, hydroxy-5 indole, 5-hydroxy-indole, 5-indolol, hydroxy-5-indole, 5-hydroxyindol, 5-hydroxy indole, unii-320un7xzyn
PubChem CID 16054
IUPAC Name 1H-indol-5-ol
SMILES OC1=CC=C2NC=CC2=C1

Specifications

Melting Point 106°C to 111°C
Quantity 1 g
Beilstein 112349
Formula Weight 133.15
Percent Purity ≥98%
Chemical Name or Material 5-Hydroxyindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.