missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-acetic acid, MedChemExpress

Product Code. 16409336
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16409336 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16409336 Supplier Medchem Express Supplier No. HYW008253
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress 5-Hydroxyindole-3-acetic acid is the main metabolite of serotonin or metanephrines, which can be used as a biomarker of neuroendocrine tumors.

5-Hydroxyindole-3-acetic acid is the main metabolite of serotonin or metanephrines, which can be used as a biomarker of neuroendocrine tumors.

Specifications

Chemical Name or Material 5-Hydroxyindole-3-acetic acid
CAS 54-16-0
Color Light Nude
Purity Grade Notes Research
Molecular Formula C10H9NO3
Quantity 100 mg
Solubility Information Ethanol : 50 mg/mL (261.53 mM; Need ultrasonic)
SMILES O=C(O)CC1=CNC2=C1C=C(O)C=C2
Molecular Weight (g/mol) 191.18
Formula Weight 191.18
Percent Purity 95.9%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.