missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-acetic Acid, 99%

Product Code. 10793861 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10793861 1 g Glass bottle 1g
10235160 250 mg Glass bottle 250mg
10524074 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10793861 Supplier Thermo Scientific Acros Supplier No. 121280010
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 54-16-0 | C10H9NO3 | 191.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-16-0
Molecular Formula C10H9NO3
Molecular Weight (g/mol) 191.19
MDL Number MFCD00005639
InChI Key DUUGKQCEGZLZNO-UHFFFAOYSA-N
Synonym 5-hydroxyindole-3-acetic acid, 5-hiaa, 2-5-hydroxy-1h-indol-3-yl acetic acid, 5-hydroxyindoleacetic acid, 5-hydroxyheteroauxin, 5-hydroxy-1h-indol-3-yl acetic acid, 5-oxyindoleacetic acid, hydroxyindoleacetic acid, 5-hydroxy-iaa, 5-hydroxyindol-3-ylacetic acid
PubChem CID 1826
ChEBI CHEBI:27823
IUPAC Name 2-(5-hydroxy-1H-indol-3-yl)acetic acid
SMILES C1=CC2=C(C=C1O)C(=CN2)CC(=O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 162°C to 165°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: slightly soluble
Formula Weight 191.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyindole-3-acetic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.