missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-acetic Acid(>90%), TRC

Product Code. 30136431
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136431 500 mg 500mg
17929354 100 mg 100mg
17907734 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136431 Supplier Toronto Research Chemicals Supplier No. TRCH953605500MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54-16-0
Molecular Formula C10 H9 N O3
Molecular Weight (g/mol) 191.18
Synonym 1H-Indole-3-acetic acid, 5-hydroxy-, Indole-3-acetic acid, 5-hydroxy- (8CI), (5-Hydroxy-1H-indol-3-yl)acetic acid, 2-(5-Hydroxy-1H-indol-3-yl)acetic acid, 5-HIAA, 5-Hydroxy-1H-indole-3-acetic acid, 5-Hydroxy-3-indolylacetate, 5-Hydroxy-IAA, 5-Hydroxyheteroauxin, 5-Hydroxyindol-3-ylacetic acid, 5-Hydroxyindole-3-acetic acid, 5-Hydroxyindoleacetic acid, 5-Oxyindoleacetic acid, NSC 90432
IUPAC Name 2-(5-hydroxy-1H-indol-3-yl)acetic acid
SMILES OC(=O)Cc1c[nH]c2ccc(O)cc12

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H9NO3/c12-7-1-2-9-8(4-7)6(5-11-9)3-10(13)14/h1-2,4-5,11-12H,3H2,(H,13,14)
Formula Weight 191.0582
Percent Purity >95%
Chemical Name or Material 5-Hydroxyindolyl-3-Acetic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.