missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyindole-3-acetic Acid-13C3, TRC

Product Code. 30394598
Change view
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30394598 0.5 mg 0.5mg
30398174 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30394598 Supplier Toronto Research Chemicals Supplier No. TRCH9536040.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Hydroxyindole-3-acetic Acid-13C3
Molecular Formula C713C3H9NO3
Quantity 0.5 mg
Synonym (5-Hydroxy-1H-indol-3-yl)acetic Acid-13C3, 5-HIAA-13C3, 5-Hydroxy-1H-indole-3-acetic Acid-13C3, 5-Hydroxy-3-indolylacetate-13C3, 5-Hydroxy-IAA-13C3, 5-Hydroxyheteroauxin-13C3, 5-Hydroxyindol-3-ylacetic Acid-13C3, 5-Hydroxyindole-3-acetic Acid-13C3, 5-Hydroxyindoleacetic Acid-13C3, 5-Oxyindoleacetic Acid-13C3, NSC 90432-13C3
InChI Formula InChI=1S/C10H9NO3/c12-7-1-2-9-8(4-7)6(5-11-9)3-10(13)14/h1-2,4-5,11-12H,3H2,(H,13,14)/i3+1,5+1,10+1
SMILES OC1=CC=C2C(C([13CH2][13C](O)=O)=[13CH]N2)=C1
Molecular Weight (g/mol) 194.16
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.