missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxydecanoic acid sodium salt, 98%

Product Code. 15484039
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15484039 500 mg 500mg
15474039 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15484039 Supplier Thermo Scientific Alfa Aesar Supplier No. J61434.ME
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 624-00-0 | C10H20O3 | 188.267 g/mol

A potassium channel antagonist which blocks the postischemic effects of the potassium channel activator cromakalim

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A potassium channel antagonist which blocks the postischemic effects of the potassium channel activator cromakalim.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-00-0
Molecular Formula C10H20O3
Molecular Weight (g/mol) 188.267
MDL Number MFCD00153808
InChI Key LMHJFKYQYDSOQO-UHFFFAOYSA-N
Synonym 5-hydroxydecanoate, 5-hydroxy capric acid, 5-hydroxy-decanoic acid, decanoic acid, 5-hydroxy, sodium salt, decanoic acid, 5-hydroxy-, 5s, acmc-20mvfk, xi-5-hydroxydecanoic acid
PubChem CID 1825
IUPAC Name 5-hydroxydecanoic acid
SMILES CCCCCC(CCCC(=O)O)O

Specifications

Color White
Quantity 500 mg
Formula Weight 210.25
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 5-Hydroxydecanoic acid sodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.