missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyanthranilic acid, 98%

Product Code. p-144402 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10149793

Brand: Thermo Scientific Acros 310310010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 394-31-0 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-31-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007870
InChI Key HYNQTSZBTIOFKH-UHFFFAOYSA-N
Synonym 5-hydroxyanthranilic acid, diabeton, benzoic acid, 2-amino-5-hydroxy, 2-amino-5-hydroxy benzoic acid, unii-uf9ww3i410, 2-amino-5-hydroxy-benzoic acid, 5-hydroxyanthranillicacid, pubchem15307, 4-amino-3-carboxyphenol, 5-hydroxyanthranillic acid
PubChem CID 164592
IUPAC Name 2-amino-5-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 233°C to 234°C
Color Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H3(NH2)CO2H
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 153.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyanthranilic acid
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.