missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyanthranilic acid, 98%

Product Code. 10703351 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10703351 10 g Glass bottle 10g
10149793 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10703351 Supplier Thermo Scientific Acros Supplier No. 310310100
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 394-31-0 | C7H7NO3 | 153.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-31-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007870
InChI Key HYNQTSZBTIOFKH-UHFFFAOYSA-N
Synonym 5-hydroxyanthranilic acid, diabeton, benzoic acid, 2-amino-5-hydroxy, 2-amino-5-hydroxy benzoic acid, unii-uf9ww3i410, 2-amino-5-hydroxy-benzoic acid, 5-hydroxyanthranillicacid, pubchem15307, 4-amino-3-carboxyphenol, 5-hydroxyanthranillic acid
PubChem CID 164592
IUPAC Name 2-amino-5-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 233°C to 234°C
Color Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H3(NH2)CO2H
Quantity 10 g
Solubility Information Solubility in water: insoluble
Formula Weight 153.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyanthranilic acid
Show More Show Less
compliance-icons
Product Identifier
  • 5-Hydroxyanthranilic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 5-Hydroxyanthranilicacid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.