missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy Rosiglitazone-d4, TRC

Product Code. 30399858
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30399858 10 mg
30406687 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30399858 Supplier Toronto Research Chemicals Supplier No. TRCH95344210MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1246817-46-8
Molecular Formula C18H15D4N3O4S
Molecular Weight (g/mol) 377.45
Synonym 5-[[4-[2-[(5-Hydroxy-2-pyridinyl)methylamino]ethoxy-d4]phenylmethyl]-2,4-thiazolidinedione
SMILES O=C(NC(S1)=O)C1CC(C=C2)=CC=C2OC([2H])([2H])C([2H])([2H])N(C)C3=NC=C(O)C=C3

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C18H19N3O4S/c1-21(16-7-4-13(22)11-19-16)8-9-25-14-5-2-12(3-6-14)10-15-17(23)20-18(24)26-15/h2-7,11,15,22H,8-10H2,1H3,(H,20,23,24)/i8D2,9D2
Formula Weight 377.1347
Chemical Name or Material 5-Hydroxy Rosiglitazone-d4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.