missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy Lansoprazole Potassium Salt, TRC

Product Code. 30011236
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30011236 50 mg 50mg
30005690 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30011236 Supplier Toronto Research Chemicals Supplier No. TRCH94371150MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1329613-29-7
Molecular Formula C16 H13 F3 N3 O3 S . K
Molecular Weight (g/mol) 423.45
Synonym 1H-Benzimidazol-6-ol, 2-[[[3-methyl-4-(2,2,2-trifluoroethoxy)-2-pyridinyl]methyl]sulfinyl]-, potassium salt (1:1), 5-Hydroxylansoprazole Potassium Salt
IUPAC Name potassium;2-[[3-methyl-4-(2,2,2-trifluoroethoxy)pyridin-2-yl]methylsulfinyl]-1H-benzimidazol-5-olate
SMILES [K+].Cc1c(CS(=O)c2nc3cc([O-])ccc3[nH]2)nccc1OCC(F)(F)F

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H14F3N3O3S.K/c1-9-13(20-5-4-14(9)25-8-16(17,18)19)7-26(24)15-21-11-3-2-10(23)6-12(11)22-15;/h2-6,23H,7-8H2,1H3,(H,21,22);/q;+1/p-1
Formula Weight 423.0267
Chemical Name or Material 5-Hydroxy Lansoprazole potassium salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.