missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy Buspirone, TRC

Product Code. 30134781
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134781 10 mg 10mg
30133500 5 mg 5mg
30005438 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134781 Supplier Toronto Research Chemicals Supplier No. TRCH83079510MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105496-33-1
Molecular Formula C21 H31 N5 O3
Molecular Weight (g/mol) 401.5
Synonym 8-[4-[4-(5-hydroxy-2-pyrimidinyl)-1-piperazinyl]butyl]-8-Azaspiro[4.5]decane-7,9-dione, 5-Hydroxybuspirone
IUPAC Name 8-[4-[4-(5-hydroxypyrimidin-2-yl)piperazin-1-yl]butyl]-8-azaspiro[4.5]decane-7,9-dione
SMILES Oc1cnc(nc1)N2CCN(CCCCN3C(=O)CC4(CCCC4)CC3=O)CC2

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C21H31N5O3/c27-17-15-22-20(23-16-17)25-11-9-24(10-12-25)7-3-4-8-26-18(28)13-21(14-19(26)29)5-1-2-6-21/h15-16,27H,1-14H2
Formula Weight 401.2427
Chemical Name or Material 5-hydroxy Buspirone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.