missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-6-methoxy Duloxetine Sulfate, TRC

Product Code. 30392273
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30392273 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30392273 Supplier Toronto Research Chemicals Supplier No. TRCH94626010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Hydroxy-6-methoxy Duloxetine Sulfate
CAS 732983-57-2
Molecular Formula C19 H21 N O6 S2
Quantity 10 mg
Synonym 5-Hydroxy-6-methoxyduloxetine Sulfate Ester, 1-Naphthalenol, 2-methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-, 1-(hydrogen sulfate), 1-Naphthalenol, 2-methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-, hydrogen sulfate (ester) (9CI), 2-Methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-1-naphthalenyl hydrogen sulfate, 5-Hydroxy-6-methoxyduloxetine sulfate, 5-Hydroxy-6-methoxyduloxetine sulfate ester, 2-Methoxy-5-[(1S)-3-(methylamino)-1-(thiophen-2-yl)propoxy]naphthalen-1-ol 1-(hydrogen sulfate), 2-Methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-1-naphthalenol 1-(Hydrogen Sulfate), LY 581920
InChI Formula InChI=1S/C19H21NO6S2/c1-20-11-10-16(18-7-4-12-27-18)25-15-6-3-5-14-13(15)8-9-17(24-2)19(14)26-28(21,22)23/h3-9,12,16,20H,10-11H2,1-2H3,(H,21,22,23)/t16-/m0/s1
SMILES CNCC[C@H](Oc1cccc2c(OS(=O)(=O)O)c(OC)ccc12)c3cccs3
IUPAC Name [2-methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-1-naphthyl] hydrogen sulfate
Molecular Weight (g/mol) 423.5
Formula Weight 423.081
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.