missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Hydroxy-6-methoxy Duloxetine Sulfate, TRC
Specifications
Specifications
| Chemical Name or Material | 5-Hydroxy-6-methoxy Duloxetine Sulfate |
| CAS | 732983-57-2 |
| Molecular Formula | C19 H21 N O6 S2 |
| Quantity | 1 mg |
| Synonym | 5-Hydroxy-6-methoxyduloxetine Sulfate Ester, 1-Naphthalenol, 2-methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-, 1-(hydrogen sulfate), 1-Naphthalenol, 2-methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-, hydrogen sulfate (ester) (9CI), 2-Methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-1-naphthalenyl hydrogen sulfate, 5-Hydroxy-6-methoxyduloxetine sulfate, 5-Hydroxy-6-methoxyduloxetine sulfate ester, 2-Methoxy-5-[(1S)-3-(methylamino)-1-(thiophen-2-yl)propoxy]naphthalen-1-ol 1-(hydrogen sulfate), 2-Methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-1-naphthalenol 1-(Hydrogen Sulfate), LY 581920 |
| InChI Formula | InChI=1S/C19H21NO6S2/c1-20-11-10-16(18-7-4-12-27-18)25-15-6-3-5-14-13(15)8-9-17(24-2)19(14)26-28(21,22)23/h3-9,12,16,20H,10-11H2,1-2H3,(H,21,22,23)/t16-/m0/s1 |
| SMILES | CNCC[C@H](Oc1cccc2c(OS(=O)(=O)O)c(OC)ccc12)c3cccs3 |
| IUPAC Name | [2-methoxy-5-[(1S)-3-(methylamino)-1-(2-thienyl)propoxy]-1-naphthyl] hydrogen sulfate |
| Molecular Weight (g/mol) | 423.5 |
| Formula Weight | 423.081 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?