missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-3-phenylbenzoic Acid, TRC

Product Code. 30393273
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393273 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30393273 Supplier Toronto Research Chemicals Supplier No. TRCH95295825MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5-Hydroxy-3-phenylbenzoic Acid
CAS 35489-88-4
Molecular Formula C13H10O3
Quantity 25 mg
Synonym 5-Hydroxy-[1,1'-biphenyl]-3-carboxylic Acid
InChI Formula InChI=1S/C13H10O3/c14-12-7-10(6-11(8-12)13(15)16)9-4-2-1-3-5-9/h1-8,14H,(H,15,16)
SMILES OC(=O)c1cc(O)cc(c1)c2ccccc2
IUPAC Name 3-hydroxy-5-phenylbenzoic acid
Molecular Weight (g/mol) 214.22
Formula Weight 214.063
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.