missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Product Code. p-7030857
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15484357

Brand: Thermo Scientific Alfa Aesar H31734.MD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 216766-12-0
Molecular Formula C6H4F3NO
Molecular Weight (g/mol) 163.10
MDL Number MFCD06801325
InChI Key VFOBDHYPESAMAF-UHFFFAOYSA-N
Synonym 6-trifluoromethyl pyridin-3-ol, 5-hydroxy-2-trifluoromethyl pyridine, 6-trifluoromethyl-3-pyridinol, 2-trifluoromethyl pyridin-5-ol, 5-hydroxy-2-trifluoromethylpyridine, 3-pyridinol, 6-trifluoromethyl, 6-trifluoromethyl-pyridin-3-ol, pubchem21818, 6-trifluoromethyl 3-pyridinol
PubChem CID 2783388
IUPAC Name 6-(trifluoromethyl)pyridin-3-ol
SMILES OC1=CN=C(C=C1)C(F)(F)F

Specifications

Melting Point 174°C to 176°C
Quantity 250 mg
Formula Weight 163.1
Percent Purity 97%
Chemical Name or Material 5-Hydroxy-2-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.