missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-nitrobenzaldehyde, 98%

Product Code. 15591835 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
10g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15591835 1 g 10g
15575036 5 g 5g
15524383 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15591835 Supplier Thermo Scientific Acros Supplier No. 121650010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 42454-06-8 | C7H5NO4 | 167.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42454-06-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007332
InChI Key XLYPHUGUKGMURE-UHFFFAOYSA-N
Synonym 3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
PubChem CID 39211
IUPAC Name 5-hydroxy-2-nitrobenzaldehyde
SMILES OC1=CC=C(C(C=O)=C1)[N+]([O-])=O

Specifications

Melting Point 168.0°C to 170.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula HOC6H3(NO2)CHO
Quantity 1 g
Beilstein 08,63
Formula Weight 167.12
Percent Purity 98%
Chemical Name or Material 5-Hydroxy-2-nitrobenzaldehyde
compliance-icons
Product Identifier
  • 5-Hydroxy-2-nitrobenzaldehyde, 98%
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.