missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-nitrobenzaldehyde, 97%, Thermo Scientific Chemicals

Product Code. 11388585
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11388585 5 g 5g
11378585 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11388585 Supplier Thermo Scientific Alfa Aesar Supplier No. L12051.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 42454-06-8 | Molecular Formula: C7H5NO4 | Molecular Weight: 167.12 g/mol

5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Hydroxy-2-nitrobenzaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Soluble in methanol.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with air, oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 42454-06-8
Molecular Formula C7H5NO4
Molecular Weight (g/mol) 167.12
MDL Number MFCD00007332
InChI Key XLYPHUGUKGMURE-UHFFFAOYSA-N
Synonym 3-formyl-4-nitrophenol, 6-nitro-3-hydroxybenzaldehyde, 2-nitro-5-hydroxybenzaldehyde, 5-hydroxy-2-nitro-benzaldehyde, benzaldehyde, 5-hydroxy-2-nitro, unii-va926z93if, 6-nitro-3-hydroxy benzaldhyde, pubchem8205, acmc-1an3l, 4-08-00-00250 beilstein handbook reference
PubChem CID 39211
IUPAC Name 5-hydroxy-2-nitrobenzaldehyde
SMILES OC1=CC=C(C(C=O)=C1)[N+]([O-])=O

Specifications

Melting Point 166°C to 168°C
Quantity 5 g
Beilstein 1910196
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 167.12
Percent Purity 97%
Chemical Name or Material 5-Hydroxy-2-nitrobenzaldehyde

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.