missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-methylpyridine, 99%

Product Code. 11441617
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441617 100 g 100g
11421617 5 g 5g
11431617 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11441617 Supplier Thermo Scientific Alfa Aesar Supplier No. B25014.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 1121-78-4 | C6H7NO | 109.128 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1121-78-4
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.128
MDL Number MFCD00006339
InChI Key DHLUJPLHLZJUBW-UHFFFAOYSA-N
Synonym 5-hydroxy-2-methylpyridine, 2-methyl-5-hydroxypyridine, 3-hydroxy-6-methylpyridine, 6-methyl-3-pyridinol, 3-pyridinol, 6-methyl, 6-methyl-3-hydroxypyridine, 5-hydroxy-2-picoline, unii-uke16jp1jr, 2-methyl-5-pyridinol, uke16jp1jr
PubChem CID 14275
IUPAC Name 6-methylpyridin-3-ol
SMILES CC1=NC=C(C=C1)O

Specifications

Melting Point 167°C to 171°C
Odor Odorless
Quantity 100 g
Beilstein 107077
Formula Weight 109.13
Percent Purity 99%
Chemical Name or Material 5-Hydroxy-2-methylpyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.