missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxy-2-methylbenzoic acid, 97%, Thermo Scientific™

Product Code. 15407278
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 15407278

Brand: Thermo Scientific Alfa Aesar H60774.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Hydroxy-2-methylbenzoic acid is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Keep container tightly sealed. Store in refrigerated conditions. Incompatible wit
TRUSTED_SUSTAINABILITY

Specifications

CAS 578-22-3
Melting Point 182°C to 187°C
Molecular Formula C8H8O3
MDL Number MFCD09833184
Quantity 1 g
Synonym 5-hydroxy-2-methyl-benzoic acid, 5,2-cresotic acid, 5-hydroxy-o-toluic acid, benzoic acid, 5-hydroxy-2-methyl, 2-methyl-5-hydroxybenzoic acid, acmc-1b0c9, ksc494s9h, 5-hydroxy-2-methylbenzoic acid
Solubility Information Slightly soluble in water.
InChI Key ZIOYQUNKXJQXQY-UHFFFAOYSA-N
SMILES CC1=CC=C(O)C=C1C(O)=O
IUPAC Name 5-hydroxy-2-methylbenzoic acid
Molecular Weight (g/mol) 152.15
PubChem CID 235188
Formula Weight 152.15
Percent Purity 97%
Chemical Name or Material 5-Hydroxy-2-methylbenzoic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.